Learn More
Abcam 10 x RIPA Buffer, 15ML
Catalog No. NC0582350
Encompass_Preferred
10X RIPA lysis buffer (pH 7.5) is a means of cell lysis and protein solubilization for both adherent and suspension-cultured mammalian cells. - Cited in over 180 publications - Suitable for immunoprecipitation (IP), ELISA, SDS-PAGE, western blot - Effectively extracts cytoplasmic, nuclear and membrane proteins
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0582350 Supplier Abcam Supplier No. AB15603415ML
10X RIPA lysis buffer (pH 7.5) is a means of cell lysis and protein solubilization for both adherent and suspension-cultured mammalian cells. - Cited in over 180 publications - Suitable for immunoprecipitation (IP), ELISA, SDS-PAGE, western blot - Effectively extracts cytoplasmic, nuclear and membrane proteins
The product is subject to the following: Abcam Restricted Use Statement