Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Tris Glycine Transfer Buffer (10X) (Transblotting Buffer 10X), 1000 mL
SDP

Catalog No. 507517374
Encompass_Preferred

Tris Glycine Transfer Buffer at a 1X concentration with the addition of 20% methanol is used for electrophoretic transfer of proteins from denaturing polyacrylamide gel to a solid membrane i.e. Western blotting or Tris-glycine gel electrophoresis.

Storage Conditions: 15 °C to 30 °C

Limit exposure to light by wrapping in foil or storing in a cabinet

Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase and Protease Tested
Max Shelf Life: 24 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7374 Supplier Quality Biological Inc Supplier No. 351087131
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tris Glycine Transfer Buffer at a 1X concentration with the addition of 20% methanol is used for electrophoretic transfer of proteins from denaturing polyacrylamide gel to a solid membrane i.e. Western blotting or Tris-glycine gel electrophoresis.

Storage Conditions: 15 °C to 30 °C

Limit exposure to light by wrapping in foil or storing in a cabinet

Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase and Protease Tested
Max Shelf Life: 24 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.