Learn More
AdipoGen 10Z-Hymenialdisine (500 µg)
SDP
Supplier:  AdipoGen AGCN20067C500
Encompass_Preferred
Chemical. CAS: 82005-12-7. Formula: C11H10BrN5O2. MW: 324.1. Isolated from sponge Axinella carteri. Insecticidal and cytotoxic. Potent NF-kappaB inhibitor. Inhibits various pro-inflammatory cytokines such as IL-1, IL-2, IL-6, IL-8, TNF-alpha and nitric oxide (NO) in a variety of cell lines. ATP-competitive kinase inhibitor. Inhibits DNA damage checkpoint at G2, cyclin-dependent kinases CDK1/cyclin B, CDK2/cyclin A, CDK2/cyclin E, CDK4/cyclin D1, CDK5/p25, GSK-3beta and casein kinase 1 (CK1). Potent mitogen-activated protein kinase kinase-1 (MEK-1) inhibitor. MARK (microtubule affinity-regulating kinase) inhibitor.
Catalog No. 50-205-3413