Learn More
MSC Value Collection ZMSCH70011 5/16" ID x 7/16" OD Dash 011 Buna-N O-Ring 1/16" Thick, Round Cross Section, Shore 65 - 75, -30 to 250°F
Catalog No. 501111742
Encompass
Value Collection ZMSCH70011 5/16" ID x 7/16" OD Dash 011 Buna-N O-Ring 1/16" Thick, Round Cross Section, Shore 65 - 75, -30 to 250°F
Catalog No. 50-111-1742 Supplier MSC Supplier No. 09260118
Value Collection ZMSCH70011 5/16" ID x 7/16" OD Dash 011 Buna-N O-Ring 1/16" Thick, Round Cross Section, Shore 65 - 75, -30 to 250°F