Learn More
Cayman Chemical 11 12-DIHET 100UG
SDP
Catalog No. 502896794
Encompass_Preferred
The diol resulting from the sEH opening of (±)11(12)-EET; contracts artery rings with approximately 70% of the magnitude of (±)11(12)-EET
Catalog No. 50-289-6794 Supplier Cayman Chemical Supplier No. 51511100 UG
The diol resulting from the sEH opening of (±)11(12)-EET; contracts artery rings with approximately 70% of the magnitude of (±)11(12)-EET