Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

11,12-Dihydroindolo[2,3-a]carbazole 98.0+%, TCI America™
SDP

Catalog No. D4803200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4803200MG 200 mg
D48031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4803200MG Supplier TCI America Supplier No. D4803200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60511-85-5
Molecular Formula C18H12N2
MDL Number MFCD07371389

Specifications

Melting Point 371°C
Color White-Yellow
Quantity 200 mg
Formula Weight 256.31
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 11,12-Dihydroindolo[2,3-a]carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.