Learn More
Cayman Chemical 11 12-EET methyl ester 100UG
SDP
Catalog No. 502896466
Encompass_Preferred
A more stable form of (±)11(12)-EET used for long-term storage
Catalog No. 50-289-6466 Supplier Cayman Chemical Supplier No. 50510100 UG
A more stable form of (±)11(12)-EET used for long-term storage