Learn More
Cayman Chemical 11 12DiHETd11 GC MS Standards
SDP
Catalog No. 502889886
Encompass_Preferred
An internal standard for the quantification of 11(12)-DiHET by GC- or LC-MS
Catalog No. 50-288-9886 Supplier Cayman Chemical Supplier No. 1000797550 UG
An internal standard for the quantification of 11(12)-DiHET by GC- or LC-MS