Learn More
Cayman Chemical 11 12EETd11 GC MS Standards
SDP
Catalog No. 502890589
Encompass_Preferred
An internal standard for the quantification of 11(12)-EET by GC- or LC-MS
Catalog No. 50-289-0589 Supplier Cayman Chemical Supplier No. 10006413100 UG
An internal standard for the quantification of 11(12)-EET by GC- or LC-MS