Learn More
Cayman Chemical 11 12EETd11 GC MS Standards
SDP
Supplier:  Cayman Chemical 1000641310 UG
Encompass_Preferred
An internal standard for the quantification of 11(12)-EET by GC- or LC-MS
Catalog No. 50-288-7891