Learn More
Cayman Chemical 11 12EETd11 methyl este 100ug
SDP
Catalog No. 502990202
Encompass_Preferred
An internal standard for the quantification of 11(12)-EET methyl ester by GC- or LC-MS
Catalog No. 50-299-0202 Supplier Cayman Chemical Supplier No. 29401100 UG
An internal standard for the quantification of 11(12)-EET methyl ester by GC- or LC-MS