Learn More
Cayman Chemical 11 12EpETE Fatty Acids and Der
Supplier: Cayman Chemical 1046225 UG
An epoxygenase pathway product produced from EPA by CYP450 both in vitro and in vivo; its effects of remain to be determined