Learn More
Cayman Chemical 11 12EpETE Fatty Acids and Der
SDP
Catalog No. 502891421
Encompass_Preferred
An epoxygenase pathway product produced from EPA by CYP450 both in vitro and in vivo; its effects of remain to be determined
Catalog No. 50-289-1421 Supplier Cayman Chemical Supplier No. 10462100 UG
An epoxygenase pathway product produced from EPA by CYP450 both in vitro and in vivo; its effects of remain to be determined