Learn More
Cayman Chemical 11 12EpETE Fatty Acids and Der
SDP
Supplier:  Cayman Chemical 1046250 UG
Encompass_Preferred
An epoxygenase pathway product produced from EPA by CYP450 both in vitro and in vivo; its effects of remain to be determined
Catalog No. 50-288-7972