Learn More
Medchemexpress LLC 11,14,20,23-tetraoxa-2,8,17,26,31-pentaazanonatetracontanedioic acid, 3-carboxy-30-[(1,1-dim | 1662688-20-1 | MFCD33551739 | 99.7% | 1196.51 g/mol | C64H101N5O16 | 100 MG
SDP
Catalog No. 5000284838
Encompass_Preferred
Fmoc-L-Lys[Oct-(OtBu)-Glu-(OtBu)-AEEA-AEEA]-OH is an Fmoc-protected lysine derivative used as a building block in peptide synthesis. It features an octanoyl-modified lysine side chain, tert-butyl-protected glutamate, and two AEEA spacers to enable incorporation of lipidated or spacer-linked functionalities into peptide sequences. Intended for research use only; follow supplier handling and storage recommendations.
Catalog No. 50-002-84838 Supplier Medchemexpress LLC Supplier No. HYW615153100MG
Fmoc-L-Lys[Oct-(OtBu)-Glu-(OtBu)-AEEA-AEEA]-OH is an Fmoc-protected lysine derivative used as a building block in peptide synthesis. It features an octanoyl-modified lysine side chain, tert-butyl-protected glutamate, and two AEEA spacers to enable incorporation of lipidated or spacer-linked functionalities into peptide sequences. Intended for research use only; follow supplier handling and storage recommendations.