Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 11,15-O-dimethyl-PGE2 | 211230-67-0 | 98.3% | 50 MG
SDP

Catalog No. 5000231036
Encompass_Preferred

ONO-AE 248 is a selective EP3 prostaglandin receptor agonist used in preclinical pharmacology and cardiovascular research. It is reported to reduce myocardial infarction size and is used as a research reagent.

  • Selective EP3 receptor agonist used in mechanism and pharmacology studies.
  • Reported cardioprotective activity in preclinical models.
  • High reported purity (98.3%) suitable for research applications.
  • Molecular formula C22H36O5; molecular weight 380.52 g/mol.
  • Soluble in DMSO; store protected from light at 4°C.

Catalog No. 50-002-31036 Supplier Medchemexpress LLC Supplier No. HY1218450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ONO-AE 248 is a selective EP3 prostaglandin receptor agonist used in preclinical pharmacology and cardiovascular research. It is reported to reduce myocardial infarction size and is used as a research reagent.

  • Selective EP3 receptor agonist used in mechanism and pharmacology studies.
  • Reported cardioprotective activity in preclinical models.
  • High reported purity (98.3%) suitable for research applications.
  • Molecular formula C22H36O5; molecular weight 380.52 g/mol.
  • Soluble in DMSO; store protected from light at 4°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.