Learn More
Medchemexpress LLC 11-{[2-(2,6-dioxopiperidin-3-yl)-1,3-dioxo-2,3-dihydro-1H-isoindol-4-yl]oxy}undecanoic acid | 2379870-45-6 | MFCD03424822 | 99.1% | C24H30N2O7 | 10MG
SDP
Catalog No. 5000259722
Encompass_Preferred
Thalidomide-4-O-C10-COOH is a thalidomide-derived cereblon (CRBN) E3 ligase ligand-linker conjugate provided for research use as a synthetic intermediate in the preparation of proteolysis-targeting chimeras (PROTACs). The molecule features a ten-carbon linker terminating in a carboxylic acid, and is supplied as a solid with formula C24H30N2O7 and molecular weight 458.50 g·mol⁻¹.
  • Thalidomide-based cereblon (CRBN) ligand-linker conjugate.
  • Suitable for PROTAC synthesis and E3 ligase recruitment studies.
  • Contains a C10 linker with a terminal carboxylic acid for conjugation.
  • High reported purity (99.1%).
  • Supplied as a solid; molecular formula C24H30N2O7, molecular weight 458.50 g·mol⁻¹.
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-59722 Supplier Medchemexpress LLC Supplier No. HY13285810MG
Thalidomide-4-O-C10-COOH is a thalidomide-derived cereblon (CRBN) E3 ligase ligand-linker conjugate provided for research use as a synthetic intermediate in the preparation of proteolysis-targeting chimeras (PROTACs). The molecule features a ten-carbon linker terminating in a carboxylic acid, and is supplied as a solid with formula C24H30N2O7 and molecular weight 458.50 g·mol⁻¹.
  • Thalidomide-based cereblon (CRBN) ligand-linker conjugate.
  • Suitable for PROTAC synthesis and E3 ligase recruitment studies.
  • Contains a C10 linker with a terminal carboxylic acid for conjugation.
  • High reported purity (99.1%).
  • Supplied as a solid; molecular formula C24H30N2O7, molecular weight 458.50 g·mol⁻¹.
  • For research use only; not for human therapeutic use.