Learn More
Medchemexpress LLC 11-(2-fluoropyridin-4-yl)-4,8,10-triazatricyclo[7.4.0.02,7]trideca-1(9),2(7),3,5,10,12-hexaene | 2173361-80-1 | 99.9% | C15H9FN4 | 10MG
SDP
Catalog No. 5000260229
Encompass_Preferred
Tau tracer 2 is a small-molecule PET tracer developed for imaging aggregated tau protein in neurodegenerative disease research. It binds selectively to tau aggregates and is used in preclinical and diagnostic studies to assess regional tau pathology with PET imaging.
  • High affinity for aggregated tau enabling sensitive PET imaging.
  • Suitable for preclinical and diagnostic research applications.
  • High chemical purity suitable for analytical and radiolabeling workflows.
  • Supplied in small-mass packages for tracer synthesis and testing.
  • Characterized by molecular formula C15H9FN4 and CAS 2173361-80-1.
  • Datasheet available with analytical and handling information.
Catalog No. 50-002-60229 Supplier Medchemexpress LLC Supplier No. HY13488010MG
Tau tracer 2 is a small-molecule PET tracer developed for imaging aggregated tau protein in neurodegenerative disease research. It binds selectively to tau aggregates and is used in preclinical and diagnostic studies to assess regional tau pathology with PET imaging.
  • High affinity for aggregated tau enabling sensitive PET imaging.
  • Suitable for preclinical and diagnostic research applications.
  • High chemical purity suitable for analytical and radiolabeling workflows.
  • Supplied in small-mass packages for tracer synthesis and testing.
  • Characterized by molecular formula C15H9FN4 and CAS 2173361-80-1.
  • Datasheet available with analytical and handling information.