Learn More
Chemglass Life Sciences Stopcock, Straight Bore, Glass Shell only for PTFE Plug, 1mm, 1.5mm & 2mm Bore, 11/25 Barrel Size, 8mm OD Sidearms
SDP
Catalog No. CG42603
Encompass_Preferred
Stopcock, Straight Bore, Glass Shell only for PTFE Plug, 1mm, 1.5mm & 2mm Bore, 11/25 Barrel Size, 8mm OD Sidearms. Glass portion only of CG-424 stopcock. Accepts 1:5 taper PTFE plugs.
Catalog No. CG42603 Supplier Chemglass Life Sciences Supplier No. CG42603
Stopcock, Straight Bore, Glass Shell only for PTFE Plug, 1mm, 1.5mm & 2mm Bore, 11/25 Barrel Size, 8mm OD Sidearms. Glass portion only of CG-424 stopcock. Accepts 1:5 taper PTFE plugs.