Learn More
Chemglass Life Sciences Replacement Washers for 11/25 PTFE Plug
SDP
Catalog No. 50872425
Encompass_Preferred
Replacement Washers for 11/25 PTFE Plug. Replacement components for 1:5 taper PTFE plugs. O-rings are Viton, nuts are green, and washers are white.
Catalog No. 50-872-425 Supplier Chemglass Life Sciences Supplier No. CG45702
Replacement Washers for 11/25 PTFE Plug. Replacement components for 1:5 taper PTFE plugs. O-rings are Viton, nuts are green, and washers are white.