Learn More
Chemglass Life Sciences 1, 1.5 and 2mm Bore, 11/25 Plug Size, 8mm OD Sidearm
SDP
Catalog No. 501531994
Encompass_Preferred
1, 1.5 and 2mm Bore, 11/25 Plug Size, 8mm OD Sidearm. Glass portion only of CG-424 stopcock. Accepts 1:5 taper PTFE plugs.
Catalog No. 50-153-1994 Supplier Chemglass Life Sciences Supplier No. CGB42603
1, 1.5 and 2mm Bore, 11/25 Plug Size, 8mm OD Sidearm. Glass portion only of CG-424 stopcock. Accepts 1:5 taper PTFE plugs.