Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 11-Amino-1-undecanethiol hydrochloride

Catalog No. 67439750MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-439-750MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-439-750MG Supplier Sigma Aldrich Supplier No. 67439750MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

11-Amino-1-undecanethiol hydrochloride

Specifications

CAS 143339-58-6
Melting Point 120°C to 170°C
Molecular Formula C11H25NS -+ HCl
Linear Formula HSCH2(CH2)9 CH2NH2 -+ HCl
Quantity 50 mg
MDL Number MFCD09836217
Synonym AUT; 11-Aminoundecanethiol hydrochloride; 11-Mercaptoundecylamine; MUAM
Molecular Weight (g/mol) 239.85
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.