Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 11-aminoundecanoic acid | 2432-99-7 | MFCD00008150 | 97.0% | 201.31 g/mol | C11H23NO2 | 100 G
SDP

Catalog No. 5000303896
Encompass_Preferred

11-Aminoundecanoic acid is an alkyl-chain amino acid used as a PROTAC linker and synthetic building block in medicinal chemistry and chemical biology. It functions as a flexible spacer to connect targeting ligands and E3 ligands in proteolysis-targeting chimera (PROTAC) constructs.

  • Alkyl-chain amino acid suitable as a PROTAC linker.
  • Provides a flexible, hydrophobic spacer for linker design.
  • Solid, white to off-white powder with reported purity of 97.0%.
  • Molecular formula C11H23NO2 and molecular weight 201.31 g/mol.
  • Store at room temperature for product stability; solution storage recommendations available in supplier documentation.

Catalog No. 50-003-03896 Supplier Medchemexpress LLC Supplier No. HYW014831100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

11-Aminoundecanoic acid is an alkyl-chain amino acid used as a PROTAC linker and synthetic building block in medicinal chemistry and chemical biology. It functions as a flexible spacer to connect targeting ligands and E3 ligands in proteolysis-targeting chimera (PROTAC) constructs.

  • Alkyl-chain amino acid suitable as a PROTAC linker.
  • Provides a flexible, hydrophobic spacer for linker design.
  • Solid, white to off-white powder with reported purity of 97.0%.
  • Molecular formula C11H23NO2 and molecular weight 201.31 g/mol.
  • Store at room temperature for product stability; solution storage recommendations available in supplier documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.