Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

11-Aminoundecyltriethoxysilane, 90+%

Catalog No. AAH3383503
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3383503 1 g
AAH33835MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3383503 Supplier Thermo Scientific Chemicals Supplier No. H3383503
Only null left
Add to Cart

CAS: 116821-45-5 | C17H39NO3Si | 333.588 g/mol

Used as primary and secondary intermediate for pharmaceutical and chemical additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as primary and secondary intermediate for pharmaceutical and chemical additives.

Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116821-45-5
Molecular Formula C17H39NO3Si
Molecular Weight (g/mol) 333.588
MDL Number MFCD08272792
InChI Key LPWZCJFZJCOBHO-UHFFFAOYSA-N
PubChem CID 18944844
IUPAC Name 11-triethoxysilylundecan-1-amine
SMILES CCO[Si](CCCCCCCCCCCN)(OCC)OCC

Specifications

Boiling Point 110°C to 115°C (0.02mmHg)
Quantity 1 g
Linear Formula H2N(CH2)11Si(OCH2CH3)3
Synonym 11-triethoxysilyl-1-undecanamine, 11-aminoundecyltriethoxysilane, 11-triethoxysilyl undecan-1-amine, 1-undecanamine,11-triethoxysilyl, 11-aminoundecyl triethoxysilane, acmc-20mmty, 1-undecanamine, 11-triethoxysilyl
Formula Weight 333.59
Percent Purity 95%
Sensitivity Moisture sensitive
Chemical Name or Material 11-Aminoundecyltriethoxysilane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.