Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 11-Azido-3,6,9-Trioxaundecan-1-Amine

Catalog No. 177585ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-758-5ML 5 mL
17-758-1ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 17-758-5ML Supplier Sigma Aldrich Supplier No. 177585ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

11-Azido-3,6,9-trioxaundecan-1-amine is an azide with polyethylene glycol-like characteristics that can be used to prepare azide-functionalized polymers via click reaction.Bifunctional, hydrophilic linker

Specifications

CAS 134179-38-7
Density 1.10 g/mL at 20°C
Molecular Formula C8H18N4O3
Refractive Index n20/D 1.470
Linear Formula C8H18N4O3
MDL Number MFCD00269874
Quantity 5 mL
Synonym 1-Amino-11-azido-3,6,9-trioxaundecane; 2-{2-[2-(2-Azidoethoxy)ethoxy]ethoxy}ethylamine; O-(2-Aminoethyl)-O'-(2-azidoethyl)diethylene glycol
Molecular Weight (g/mol) 218.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.