Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 11-Bromo-1-undecanethiol

Catalog No. 701335250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-133-5250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-133-5250MG Supplier Sigma Aldrich Supplier No. 701335250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

11-Bromo-1-undecanethiol (BrUDT) is a bromo-terminated alkanethiol that forms a self-assembled monolayer(SAM) on a variety of surfaces. It can modify the surface characteristics by forming a bond between the sulfur groups and surface atoms.BrUDT forms a protective SAM on gold nanoparticles, which can be potentially used in biomedical applications for the fabrication of in vivo sensors. It can also be mixed with dodecanethiol, which can be coated on gold nanoparticles for the synthesis of gasotransmitters.

Specifications

CAS 116129-34-1
Density 1.128 g/mL (at 25°C)
Molecular Formula C11H23BrS
Refractive Index n20/D 1.501
Linear Formula Br(CH2)10 CH2SH
Quantity 250 mg
MDL Number MFCD12406810
Molecular Weight (g/mol) 267.27
Percent Purity 99%
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.