Learn More
Medchemexpress LLC 11-bromo-1-undecanol | 1611-56-9 | MFCD00004752 | 99.2% | 251.20 g/mol | C11H23BrO | 10 G
SDP
Catalog No. 5000303233
Encompass_Preferred
11-bromo-1-undecanol is a brominated C11 primary alcohol used as a versatile synthetic intermediate and PROTAC linker in medicinal chemistry. It offers both a terminal bromide and a primary hydroxyl group for derivatization and conjugation in linker construction.
  • Primary bromide and alcohol functional groups for diverse derivatization.
  • Useful as a linker in PROTAC synthesis and conjugation chemistry.
  • Available in multiple quantities to support discovery and scale-up.
  • White to beige crystalline solid suitable for handling and storage.
  • Molecular formula C11H23BrO; molar mass 251.20 g/mol.
Catalog No. 50-003-03233 Supplier Medchemexpress LLC Supplier No. HYW01390710G
11-bromo-1-undecanol is a brominated C11 primary alcohol used as a versatile synthetic intermediate and PROTAC linker in medicinal chemistry. It offers both a terminal bromide and a primary hydroxyl group for derivatization and conjugation in linker construction.
  • Primary bromide and alcohol functional groups for diverse derivatization.
  • Useful as a linker in PROTAC synthesis and conjugation chemistry.
  • Available in multiple quantities to support discovery and scale-up.
  • White to beige crystalline solid suitable for handling and storage.
  • Molecular formula C11H23BrO; molar mass 251.20 g/mol.