Learn More
AdipoGen 11-Bromo-1-undecanol
SDP
Supplier:  AdipoGen CDXB0626G010
Encompass_Preferred
Chemical. CAS 1611-56-9. Formula C11H23BrO. MW 251.2. 11-Bromo-1-undecanol is a bulding block for synthesis. It was used in the preparation of graft copolymers based on a metathesis skeleton with polymethyl methacrylate grafts and in the synthesis of polymerizable tail groups consisting of n-Alkoxy-2,4-hexadiene used in the design of radically polymerized ionic liquid crystal monomers.
Catalog No. 50-256-1578