Learn More
Medchemexpress LLC 11-bromoundecan-1-ol | 1611-56-9 | MFCD00004752 | 251.20 g/mol | C11H23BrO | 25 G
SDP
Catalog No. 5000303506
Encompass_Preferred
11-Bromoundecan-1-ol is a bromoalkyl alcohol used as a PROTAC linker and synthetic intermediate in the construction of small-molecule degraders. Its terminal bromide serves as an electrophilic handle for substitution chemistry, while the 11-carbon alkyl spacer provides flexibility and hydrophobic separation between conjugated ligands. Intended for research use.
Catalog No. 50-003-03506 Supplier Medchemexpress LLC Supplier No. 25GHYW013907
11-Bromoundecan-1-ol is a bromoalkyl alcohol used as a PROTAC linker and synthetic intermediate in the construction of small-molecule degraders. Its terminal bromide serves as an electrophilic handle for substitution chemistry, while the 11-carbon alkyl spacer provides flexibility and hydrophobic separation between conjugated ligands. Intended for research use.