Learn More
Cayman Chemical 11-cIs RetInal 2mg
SDP
Catalog No. 502981514
Encompass_Preferred
An inverse agonist of opsin; isomerizes to all-trans retinal to start the phototransduction cycle of mammalian vision; binds as a partial agonist to a G121L-mutated opsin; application to compound eyes restored moth visual sensitivity almost completely
Catalog No. 50-298-1514 Supplier Cayman Chemical Supplier No. 213772 MG
An inverse agonist of opsin; isomerizes to all-trans retinal to start the phototransduction cycle of mammalian vision; binds as a partial agonist to a G121L-mutated opsin; application to compound eyes restored moth visual sensitivity almost completely