Learn More
TARGETMOL CHEMICALS INC 11-cis-Vaccenyl acetate 25MG
SDP
Catalog No. 502880555
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 11-cis-Vaccenyl acetate ((Z)-Octadec-11-enyl acetate) activates olfactory neurons located in the T1 sensilla on the antenna and mediates aggregation behavior in both male and female flies. Purity 96.53%
Catalog No. 50-288-0555 Supplier TARGETMOL CHEMICALS INC Supplier No. T1003825MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 11-cis-Vaccenyl acetate ((Z)-Octadec-11-enyl acetate) activates olfactory neurons located in the T1 sensilla on the antenna and mediates aggregation behavior in both male and female flies. Purity 96.53%