Learn More
Medchemexpress LLC 11-cis-vaccenyl acetate | 6186-98-7 | MFCD00056312 | 98.9% | 310.51 g/mol | C20H38O2 | 5 MG
SDP
Catalog No. 5000262326
Encompass_Preferred
11-cis-Vaccenyl acetate is an endogenous insect pheromone used in biochemical and behavioral research to investigate olfactory signaling and aggregation behavior, particularly in Drosophila. It is supplied as a high-purity, research-grade compound for laboratory studies.
  • High purity (98.9%) suitable for analytical and behavioral assays.
  • Chemical formula C20H38O2; molecular weight 310.51 g/mol.
  • Commonly used to study olfactory signaling and aggregation behavior in Drosophila.
  • Supplied in research-scale quantities for controlled experiments.
  • Stable when stored under recommended conditions; consult SDS for handling and safety.
Catalog No. 50-002-62326 Supplier Medchemexpress LLC Supplier No. HY1289005MG
11-cis-Vaccenyl acetate is an endogenous insect pheromone used in biochemical and behavioral research to investigate olfactory signaling and aggregation behavior, particularly in Drosophila. It is supplied as a high-purity, research-grade compound for laboratory studies.
  • High purity (98.9%) suitable for analytical and behavioral assays.
  • Chemical formula C20H38O2; molecular weight 310.51 g/mol.
  • Commonly used to study olfactory signaling and aggregation behavior in Drosophila.
  • Supplied in research-scale quantities for controlled experiments.
  • Stable when stored under recommended conditions; consult SDS for handling and safety.