Learn More
Cayman Chemical 11-dehydro Thromboxn B2 1mg
SDP
Catalog No. 502983327
Encompass_Preferred
One of the main metabolites of TXB2; can be used as a marker of in vivo TXA2 synthesis; urinary concentrations of 11-TXB2 are approximately 30-70 ng/mmol creatinine
Catalog No. 50-298-3327 Supplier Cayman Chemical Supplier No. 195001 MG
One of the main metabolites of TXB2; can be used as a marker of in vivo TXA2 synthesis; urinary concentrations of 11-TXB2 are approximately 30-70 ng/mmol creatinine