Learn More
Cayman Chemical 11-Dehydrocorticosterone, Purity: ≥90%, CAS: 72-23-1, 10 mg
SDP
Catalog No. 502950753
Encompass_Preferred
An endogenous mineralocorticoid; increases Na+/K+-ATPase mRNA expression in vascular smooth muscle cells and inhibits aldosterone action in B. marinus toad bladder tissue in a concentration-dependent manner; decreases the sodium/creatine ratio and increases the potassium/creatine ratio in rat urine in a dose-dependent manner in a model of carbenoxolone-induced 11β-hydroxysteroid dehydrogenase inhibition; chronic administration increases plasma glucocorticoids levels, body weight gain, and adiposity, as well as induces insulin resistance in mice
Catalog No. 50-295-0753 Supplier Cayman Chemical Supplier No. 3290710 MG
An endogenous mineralocorticoid; increases Na+/K+-ATPase mRNA expression in vascular smooth muscle cells and inhibits aldosterone action in B. marinus toad bladder tissue in a concentration-dependent manner; decreases the sodium/creatine ratio and increases the potassium/creatine ratio in rat urine in a dose-dependent manner in a model of carbenoxolone-induced 11β-hydroxysteroid dehydrogenase inhibition; chronic administration increases plasma glucocorticoids levels, body weight gain, and adiposity, as well as induces insulin resistance in mice