Learn More
Cayman Chemical 11-deoxy CortIcosteron 100mg
SDP
Catalog No. 502995351
Encompass_Preferred
An endogenous mineralocorticoid; a metabolite of progesterone and precursor to aldosterone and corticosterone; metabolized to the neuroactive compound THDOC, which positively modulates GABAA receptors and produces effects similar to barbiturates in rats; levels increase following naloxone or CRH injection in cynomolgus monkeys; levels decrease following dexamethasone administration; increased 3.4-fold in obese and diabetic mice
Catalog No. 50-299-5351 Supplier Cayman Chemical Supplier No. 22916100 MG
An endogenous mineralocorticoid; a metabolite of progesterone and precursor to aldosterone and corticosterone; metabolized to the neuroactive compound THDOC, which positively modulates GABAA receptors and produces effects similar to barbiturates in rats; levels increase following naloxone or CRH injection in cynomolgus monkeys; levels decrease following dexamethasone administration; increased 3.4-fold in obese and diabetic mice