Learn More
Cayman Chemical 11-deoxy Corticosterone, Purity: ≥95%, CAS: 64-85-7, 50 mg
SDP
Catalog No. 502994223
Encompass_Preferred
An endogenous mineralocorticoid; a metabolite of progesterone and precursor to aldosterone and corticosterone; metabolized to the neuroactive compound THDOC, which positively modulates GABAA receptors and produces effects similar to barbiturates in rats; levels increase following naloxone or CRH injection in cynomolgus monkeys; levels decrease following dexamethasone administration; increased 3.4-fold in obese and diabetic mice
Catalog No. 50-299-4223 Supplier Cayman Chemical Supplier No. 2291650 MG
An endogenous mineralocorticoid; a metabolite of progesterone and precursor to aldosterone and corticosterone; metabolized to the neuroactive compound THDOC, which positively modulates GABAA receptors and produces effects similar to barbiturates in rats; levels increase following naloxone or CRH injection in cynomolgus monkeys; levels decrease following dexamethasone administration; increased 3.4-fold in obese and diabetic mice