Learn More
Medchemexpress LLC 11-deoxy corticosterone-d7 | 64-85-7 | MFCD00003661 | 98.0% | 337.50 g·mol⁻1 | C21H23D7O3 | 1 MG
SDP
Catalog No. 5000284245
Encompass_Preferred
11-deoxy corticosterone-d7 is the deuterium-labeled form of 11-deoxycorticosterone used as an internal standard and tracer for quantitative analytical methods. It is supplied as a neat solid in small pack sizes for laboratory reference use.
  • Deuterium-labeled (d7) internal standard for quantitative assays.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Supplied as a neat, white to off-white solid for analytical use.
  • High isotopic and chemical purity for reliable quantitation.
  • Small pack sizes suitable for reference and tracer studies.
Catalog No. 50-002-84245 Supplier Medchemexpress LLC Supplier No. HYW6539391MG
11-deoxy corticosterone-d7 is the deuterium-labeled form of 11-deoxycorticosterone used as an internal standard and tracer for quantitative analytical methods. It is supplied as a neat solid in small pack sizes for laboratory reference use.
  • Deuterium-labeled (d7) internal standard for quantitative assays.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Supplied as a neat, white to off-white solid for analytical use.
  • High isotopic and chemical purity for reliable quantitation.
  • Small pack sizes suitable for reference and tracer studies.