Learn More
Medchemexpress LLC 11-deoxycortisol | 152-58-9 | MFCD00003662 | 98.3% | 346.46 g/mol | C21H30O4 | 10 MG
SDP
Catalog No. 5000297900
Encompass_Preferred
Cortodoxone (11-deoxycortisol) analytical standard for research and analytical applications. It is a glucocorticoid steroid reference material used in qualitative and quantitative assays, including HPLC, GC, and MS. Supplied as a white to off-white solid with high purity to support reliable method development.
  • High purity reference material for analytical assays.
  • Suitable for HPLC, GC, and MS method development.
  • White to off-white solid form for easy handling and weighing.
  • Provides consistent performance in quantitative analyses.
  • Available as a 10 mg pack for small-scale experiments.
Catalog No. 50-002-97900 Supplier Medchemexpress LLC Supplier No. HY77839R10MG
Cortodoxone (11-deoxycortisol) analytical standard for research and analytical applications. It is a glucocorticoid steroid reference material used in qualitative and quantitative assays, including HPLC, GC, and MS. Supplied as a white to off-white solid with high purity to support reliable method development.
  • High purity reference material for analytical assays.
  • Suitable for HPLC, GC, and MS method development.
  • White to off-white solid form for easy handling and weighing.
  • Provides consistent performance in quantitative analyses.
  • Available as a 10 mg pack for small-scale experiments.