Learn More
Medchemexpress LLC 11-dodecenoic acid | 65423-25-8 | MFCD00016758 | 95.0% | 198.30 g/mol | C12H22O2 | 250 MG
SDP
Catalog No. 5000281953
Encompass_Preferred
11-Dodecenoic acid is an unsaturated C12 fatty acid used as a biochemical reagent in life-science research. It is commonly employed in biochemical assays, method development, and as a reference compound, and is supplied with documented purity and recommended storage conditions.
  • Unsaturated C12 fatty acid (C12H22O2) with molecular weight 198.30 g/mol.
  • Supplied with documented purity (main listing 95.0%; batch-specific documentation may report higher values).
  • Suitable for biochemical assays, method development, and reference standard use.
  • Stable when stored under recommended conditions: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in multiple laboratory pack sizes to support small-scale research.
Catalog No. 50-002-81953 Supplier Medchemexpress LLC Supplier No. HYW320659250MG
11-Dodecenoic acid is an unsaturated C12 fatty acid used as a biochemical reagent in life-science research. It is commonly employed in biochemical assays, method development, and as a reference compound, and is supplied with documented purity and recommended storage conditions.
  • Unsaturated C12 fatty acid (C12H22O2) with molecular weight 198.30 g/mol.
  • Supplied with documented purity (main listing 95.0%; batch-specific documentation may report higher values).
  • Suitable for biochemical assays, method development, and reference standard use.
  • Stable when stored under recommended conditions: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in multiple laboratory pack sizes to support small-scale research.