Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 11-Dodecyn-1-ol | 18202-10-3 | 182.30 | 1 G
SDP

Catalog No. 5000460432
Encompass_Preferred

11-Dodecyn-1-ol is a synthetic intermediate of the sex pheromone (Z,E)-11,13,15-Hexadecatrienyl acetate for Thaumetopoea processionea. This product is for research use only.

  • Purity: 99.4%
  • CAS number: 18202-10-3
  • Molecular weight: 182.30
  • Molecular formula: C12H22O
  • Appearance: solid below 26°C, liquid above 28°C
  • Color: colorless to light yellow
  • Target: biochemical assay reagents
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility (in vitro): 200 mg/mL in DMSO

Catalog No. 50-004-60432 Supplier Medchemexpress LLC Supplier No. HYW0072731G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

11-Dodecyn-1-ol is a synthetic intermediate of the sex pheromone (Z,E)-11,13,15-Hexadecatrienyl acetate for Thaumetopoea processionea. This product is for research use only.

  • Purity: 99.4%
  • CAS number: 18202-10-3
  • Molecular weight: 182.30
  • Molecular formula: C12H22O
  • Appearance: solid below 26°C, liquid above 28°C
  • Color: colorless to light yellow
  • Target: biochemical assay reagents
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility (in vitro): 200 mg/mL in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.