Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 11-Dodecyn-1-ol | 18202-10-3 | 182.30 | 1 ML
SDP

Catalog No. 5000460406
Encompass_Preferred

11-Dodecyn-1-ol is a synthetic intermediate used in the production of sex pheromones for the oak processionary moth (*Thaumetopoea processionea*). This chemical compound is provided with a high purity and specific storage guidelines to maintain its stability and effectiveness.

  • Appears as a colorless to light yellow solid below 26°C, and a liquid above 28°C.
  • Purity of 99.40% verified by GC.
  • Can be stored in pure form at -20°C for 3 years or 4°C for 2 years.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month.
  • Soluble in DMSO at 200 mg/mL, requiring ultrasonication.
  • 1H NMR spectrum is consistent with its structure.

Catalog No. 50-004-60406 Supplier Medchemexpress LLC Supplier No. HYW00727310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

11-Dodecyn-1-ol is a synthetic intermediate used in the production of sex pheromones for the oak processionary moth (*Thaumetopoea processionea*). This chemical compound is provided with a high purity and specific storage guidelines to maintain its stability and effectiveness.

  • Appears as a colorless to light yellow solid below 26°C, and a liquid above 28°C.
  • Purity of 99.40% verified by GC.
  • Can be stored in pure form at -20°C for 3 years or 4°C for 2 years.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month.
  • Soluble in DMSO at 200 mg/mL, requiring ultrasonication.
  • 1H NMR spectrum is consistent with its structure.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.