Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 11-Dodecyn-1-ol | 18202-10-3 | 182.30 | 10 G
SDP

Catalog No. 5000460393
Encompass_Preferred

11-Dodecyn-1-ol is a synthetic intermediate of the sex pheromone (Z,E)-11,13,15-Hexadecatrienyl acetate, relevant for *Thaumetopoea processionea* (oak processionary moth).

  • Synthetic intermediate for sex pheromones
  • Appearance: colorless to light yellow (<26°C solid, >28°C liquid)
  • High purity (99.4% by GC)
  • Molecular formula C12H22O
  • Store pure form at -20°C for up to 3 years or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month
  • Soluble in DMSO at 200 mg/mL (1097.09 mM) with sonication

Catalog No. 50-004-60393 Supplier Medchemexpress LLC Supplier No. HYW00727310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

11-Dodecyn-1-ol is a synthetic intermediate of the sex pheromone (Z,E)-11,13,15-Hexadecatrienyl acetate, relevant for *Thaumetopoea processionea* (oak processionary moth).

  • Synthetic intermediate for sex pheromones
  • Appearance: colorless to light yellow (<26°C solid, >28°C liquid)
  • High purity (99.4% by GC)
  • Molecular formula C12H22O
  • Store pure form at -20°C for up to 3 years or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month
  • Soluble in DMSO at 200 mg/mL (1097.09 mM) with sonication

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.