Learn More
Cayman Chemical 11(E)-Eicosenoic Acid, Purity: ≥98%, CAS: 62322-84-3, 100 mg
SDP
Catalog No. 502986726
Encompass_Preferred
A very long-chain fatty acid; a trans monounsaturated isomer of arachidic acid; does not inhibit LTB4 (Ki = 1,150 µM); has no effect on telomerase activity
Catalog No. 50-298-6726 Supplier Cayman Chemical Supplier No. 22578100 MG
A very long-chain fatty acid; a trans monounsaturated isomer of arachidic acid; does not inhibit LTB4 (Ki = 1,150 µM); has no effect on telomerase activity