Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

11-Heneicosanone 95.0+%, TCI America™
SDP

Catalog No. H05325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05325G 5 g
H053225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05325G Supplier TCI America Supplier No. H05325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19781-72-7
Molecular Formula C21H42O
Molecular Weight (g/mol) 310.566
MDL Number MFCD00048410
InChI Key DOACSXJVHDTDSG-UHFFFAOYSA-N
Synonym Didecyl Ketone
PubChem CID 29776
IUPAC Name henicosan-11-one
SMILES CCCCCCCCCCC(=O)CCCCCCCCCC

Specifications

Melting Point 64°C
Color White-Yellow
Quantity 5 g
Formula Weight 310.57
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 11-Heneicosanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.