Learn More
Cayman Chemical 11-HexadecenoIc AcId 50mg
SDP
Catalog No. 502934602
Encompass_Preferred
A mixture of 11-cis- and 11-trans-hexadecenoic acids
Catalog No. 50-293-4602 Supplier Cayman Chemical Supplier No. 2673050 MG
A mixture of 11-cis- and 11-trans-hexadecenoic acids