Learn More
Cayman Chemical 11-hydroxy-8-THC-d3exe 100ug
SDP
Catalog No. 502944971
Encompass_Preferred
An internal standard for the quantification of (−)-11-hydroxy-Δ8-THC by GC- or LC-MS; provided as an exempt preparation; intended for research and forensic applications
Catalog No. 50-294-4971 Supplier Cayman Chemical Supplier No. 40976100 UG
An internal standard for the quantification of (−)-11-hydroxy-Δ8-THC by GC- or LC-MS; provided as an exempt preparation; intended for research and forensic applications