Learn More
Cayman Chemical 11β-Hydroxyprogesterone, Purity: ≥98%, CAS: 600-57-7, 10 mg
SDP
Catalog No. 502911978
Encompass_Preferred
A metabolite of progesterone; an MR agonist (EC50 = 0.05 µM in a reporter assay); an inhibitor of 11β-HSD2 and 11β-HSD1 (IC50s = 0.140 and 1 µM, respectively); increases blood pressure in rats at 10 µg/hour; serum levels are increased in patients with 21-hydroxylase deficiency
Catalog No. 50-291-1978 Supplier Cayman Chemical Supplier No. 3603110 MG
A metabolite of progesterone; an MR agonist (EC50 = 0.05 µM in a reporter assay); an inhibitor of 11β-HSD2 and 11β-HSD1 (IC50s = 0.140 and 1 µM, respectively); increases blood pressure in rats at 10 µg/hour; serum levels are increased in patients with 21-hydroxylase deficiency