Learn More
Medchemexpress LLC 11-Keto-beta-boswellic acid | 17019-92-0 | 470.68 | 1 ML
SDP
Catalog No. 5000415238
Encompass_Preferred
11-Keto-beta-boswellic acid is a pentacyclic triterpenic acid derived from the oleogum resin of the Boswellia serrata tree, commonly known as Indian Frankincense. It possesses anti-inflammatory activity primarily by inhibiting 5-lipoxygenase (5-LOX) and subsequently leukotriene and nuclear factor-kappa B (NF-κB) activation and tumor necrosis factor alpha generation production.
  • Anti-inflammatory activity
  • Anti-diabetic activity observed in streptozotocin-induced diabetic rats
  • Protects rats against kainic acid-induced excitotoxicity
  • Solid, white to off-white appearance
Catalog No. 50-004-15238 Supplier Medchemexpress LLC Supplier No. HYN205610MM/1ML
11-Keto-beta-boswellic acid is a pentacyclic triterpenic acid derived from the oleogum resin of the Boswellia serrata tree, commonly known as Indian Frankincense. It possesses anti-inflammatory activity primarily by inhibiting 5-lipoxygenase (5-LOX) and subsequently leukotriene and nuclear factor-kappa B (NF-κB) activation and tumor necrosis factor alpha generation production.
  • Anti-inflammatory activity
  • Anti-diabetic activity observed in streptozotocin-induced diabetic rats
  • Protects rats against kainic acid-induced excitotoxicity
  • Solid, white to off-white appearance