Learn More
Medchemexpress LLC 11-keto-beta-boswellic acid | 17019-92-0 | MFCD06656312 | 99.9% | 470.68 g/mol | C30H46O4 | 10 MG
SDP
Catalog No. 5000295473
Encompass_Preferred
11-Keto-beta-boswellic acid is a pentacyclic triterpenic acid isolated from Boswellia species with reported anti-inflammatory, metabolic, and neuroprotective activities. It modulates 5-lipoxygenase and NF-κB signaling and is commonly used in preclinical studies. Soluble in DMSO for in vitro use.
  • Pentacyclic triterpenic acid with anti-inflammatory activity.
  • Demonstrated inhibition of 5-lipoxygenase and NF-κB pathways.
  • Reported DPP-4 modulation and neuroprotective effects in animal models.
  • High purity suitable for biochemical and cell-based assays.
  • Soluble in DMSO; multiple in vivo formulation protocols reported.
Catalog No. 50-002-95473 Supplier Medchemexpress LLC Supplier No. HYN205610MG
11-Keto-beta-boswellic acid is a pentacyclic triterpenic acid isolated from Boswellia species with reported anti-inflammatory, metabolic, and neuroprotective activities. It modulates 5-lipoxygenase and NF-κB signaling and is commonly used in preclinical studies. Soluble in DMSO for in vitro use.
  • Pentacyclic triterpenic acid with anti-inflammatory activity.
  • Demonstrated inhibition of 5-lipoxygenase and NF-κB pathways.
  • Reported DPP-4 modulation and neuroprotective effects in animal models.
  • High purity suitable for biochemical and cell-based assays.
  • Soluble in DMSO; multiple in vivo formulation protocols reported.