Learn More
Cayman Chemical 11-keto Testosteron-d3 1mg
SDP
Catalog No. 502893035
Encompass_Preferred
An internal standard for the quantification of 11-keto testosterone by GC- or LC-MS
Catalog No. 50-289-3035 Supplier Cayman Chemical Supplier No. 90027541 MG
An internal standard for the quantification of 11-keto testosterone by GC- or LC-MS